Search
Search

UBE2L3| Gene Symbol: UBE2L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078094.1 | NP_001071562.1 | 3 - 4 | 348 | 88 | |
| GenBank mRNA | BC105164.1 | - | 3 - 4 | 348 | 88 | |
| BT025380.1 | - | 3 - 4 | 308 | 88 | ||
Set Membership: |
|