Search
Search
| Gene Symbol: NTS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173945.4 | NP_776370.2 | 3 - 4 | 453 | 117 | |
| GenBank mRNA | BC102381.1 | - | 3 - 4 | 454 | 117 | |
| DQ515198.1 | - | 3 - 4 | 409 | 117 | ||
| M18621.1 | - | 3 - 4 | 425 | 117 | ||
Set Membership: |
|