Search
Search

THUMPD3| Gene Symbol: THUMPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038062.1 | NP_001033151.1 | 6 - 7 | 1089 | 97 | |
| GenBank mRNA | BC111239.1 | - | 6 - 7 | 1089 | 97 | |
Set Membership: |
|