Search
Search

AZIN2| Gene Symbol: AZIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038510.3 | NP_001033599.2 | 11 - 12 | 1584 | 108 | |
| GenBank mRNA | BC109524.1 | - | 11 - 12 | 1585 | 108 | |
Set Membership: |
|