Search
Search

ATP6V1F| Gene Symbol: ATP6V1F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045869.1 | NP_001039334.1 | 1 - 2 | 246 | 78 | |
| GenBank mRNA | BC111686.1 | - | 1 - 2 | 246 | 78 | |
| U43176.1 | - | 1 - 2 | 136 | 78 | ||
Set Membership: |
|