Search
Search

SLC25A17| Gene Symbol: SLC25A17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045948.2 | NP_001039413.1 | 4 - 5 | 437 | 121 | |
| GenBank mRNA | BC105317.1 | - | 4 - 5 | 439 | 121 | |
| BT030586.1 | - | 4 - 5 | 374 | 121 | ||
Set Membership: |
|