Search
Search

HBP1| Gene Symbol: HBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046196.2 | NP_001039661.2 | 3 - 4 | 457 | 92 | |
| GenBank mRNA | BC105542.1 | - | 3 - 4 | 446 | 92 | |
| BT030675.1 | - | 2 - 3 | 422 | 92 | ||
| DV805391.1 | - | 3 - 4 | 457 | 92 | ||
| EH164246.1 | - | 3 - 4 | 436 | 92 | ||
Set Membership: |
|