Search
Search

STUB1| Gene Symbol: STUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075166.1 | NP_001068634.1 | 2 - 3 | 468 | 132 | |
| GenBank mRNA | BC109588.1 | - | 2 - 3 | 468 | 132 | |
| EU005652.1 | - | 2 - 3 | 491 | 132 | ||
Set Membership: |
|