Search
Search

CCKBR| Gene Symbol: CCKBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174262.3 | NP_776687.2 | 3 - 4 | 758 | 101 | |
| GenBank mRNA | BC112800.1 | - | 3 - 4 | 758 | 101 | |
| S83090.1 | - | 3 - 4 | 680 | 101 | ||
Set Membership: |
|