Search
Search

PON1| Gene Symbol: PON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046269.2 | NP_001039734.1 | 4 - 5 | 386 | 78 | |
| GenBank mRNA | BC112488.1 | - | 4 - 5 | 388 | 78 | |
| EU289337.1 | - | 4 - 5 | 387 | 78 | ||
Set Membership: |
|