Search
Search

C29H11orf54| Gene Symbol: C29H11orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046315.2 | NP_001039780.1 | 4 - 5 | 257 | 85 | |
| GenBank mRNA | BC105390.1 | - | 4 - 5 | 258 | 85 | |
Set Membership: |
|