Search
Search

PPA1| Gene Symbol: PPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075118.1 | NP_001068586.1 | 8 - 9 | 808 | 111 | |
| GenBank mRNA | BC114891.1 | - | 8 - 9 | 808 | 111 | |
| M95283.1 | - | 8 - 9 | 794 | 111 | ||
Set Membership: |
|