Search
Search

PCOLCE2| Gene Symbol: PCOLCE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075629.1 | NP_001069097.1 | 6 - 7 | 1088 | 77 | |
| GenBank mRNA | BC114686.1 | - | 6 - 7 | 1088 | 77 | |
Set Membership: |
|