Search
Search

NUDCD3| Gene Symbol: NUDCD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075997.1 | NP_001069465.1 | 4 - 5 | 809 | 97 | |
| GenBank mRNA | BC114136.1 | - | 4 - 5 | 809 | 97 | |
| BT021830.1 | - | 4 - 5 | 827 | 97 | ||
Set Membership: |
|