Search
Search

ABT1| Gene Symbol: ABT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076152.2 | NP_001069620.1 | 1 - 2 | 245 | 102 | |
| GenBank mRNA | BC118119.1 | - | 1 - 2 | 246 | 102 | |
| BT030713.1 | - | 1 - 2 | 235 | 102 | ||
Set Membership: |
|