Search
Search

BLCAP| Gene Symbol: BLCAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174250.3 | NP_776675.1 | 1 - 2 | 77 | 139 | |
| GenBank mRNA | AB071597.1 | - | 1 - 2 | 97 | 139 | |
| BC118229.1 | - | 1 - 2 | 77 | 139 | ||
| BT030495.1 | - | 1 - 2 | 97 | 139 | ||
Set Membership: |
|