Search
Search

IMPA1| Gene Symbol: IMPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174361.4 | NP_776786.1 | 7 - 8 | 617 | 101 | |
| GenBank mRNA | BC118411.1 | - | 7 - 8 | 618 | 101 | |
| BT030590.2 | - | 7 - 8 | 583 | 101 | ||
| J05394.1 | - | 7 - 8 | 627 | 101 | ||
Set Membership: |
|