Search
Search

UPK1B| Gene Symbol: UPK1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174482.2 | NP_776907.2 | 2 - 3 | 191 | 102 | |
| GenBank mRNA | BC116014.1 | - | 2 - 3 | 191 | 102 | |
| Z29378.1 | - | 123 | 102 | |||
Set Membership: |
|