Search
Search

SORBS1| Gene Symbol: SORBS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075169.2 | NP_001068637.2 | 15 - 16 | 1344 | 64 | |
| GenBank mRNA | BC120200.1 | - | 15 - 16 | 1345 | 64 | |
| DV820034.1 | - | 3 - 4 | 199 | 64 | ||
Set Membership: |
|