Search
Search

KCTD15| Gene Symbol: KCTD15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075568.1 | NP_001069036.1 | 3 - 4 | 333 | 71 | |
| GenBank mRNA | BC119910.1 | - | 3 - 4 | 333 | 71 | |
| BT029851.1 | - | 3 - 4 | 281 | 71 | ||
Set Membership: |
|