Search
Search

FGD2| Gene Symbol: FGD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075570.1 | NP_001069038.1 | 15 - 16 | 1913 | 123 | |
| GenBank mRNA | BT026545.1 | - | 15 - 16 | 1913 | 123 | |
Set Membership: |
|