Search
Search

DLL4| Gene Symbol: DLL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075779.1 | NP_001069247.1 | 2 - 3 | 619 | 67 | |
| GenBank mRNA | BC140546.1 | - | 2 - 3 | 546 | 67 | |
| BT026552.1 | - | 2 - 3 | 619 | 67 | ||
Set Membership: |
|