Search
Search

BPIFA2C| Gene Symbol: BPIFA2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076503.1 | NP_001069971.1 | 3 - 4 | 529 | 132 | |
| GenBank mRNA | DQ835286.1 | - | 3 - 4 | 529 | 132 | |
Set Membership: |
|