Search
Search

COL3A1| Gene Symbol: COL3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076831.1 | NP_001070299.1 | 30 - 31 | 2199 | 99 | |
| GenBank mRNA | BC123469.1 | - | 30 - 31 | 2199 | 99 | |
Set Membership: |
|