Search
Search

HNRNPLL| Gene Symbol: HNRNPLL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076900.1 | NP_001070368.1 | 12 - 13 | 1823 | 88 | |
| GenBank mRNA | BC123451.1 | - | 12 - 13 | 1823 | 88 | |
Set Membership: |
|