Search
Search

CTPS1| Gene Symbol: CTPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077858.2 | NP_001071326.1 | 11 - 12 | 1306 | 72 | |
| GenBank mRNA | BC126649.1 | - | 11 - 12 | 1308 | 72 | |
Set Membership: |
|