Search
Search

EMC10| Gene Symbol: EMC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080239.1 | NP_001073708.1 | 3 - 4 | 347 | 67 | |
| GenBank mRNA | BC123856.1 | - | 3 - 4 | 317 | 67 | |
| BC126720.1 | - | 3 - 4 | 347 | 67 | ||
Set Membership: |
|