Search
Search

JAML| Gene Symbol: JAML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080250.1 | NP_001073719.1 | 8 - 9 | 1084 | 89 | |
| GenBank mRNA | BC149327.1 | - | 7 - 8 | 1037 | 89 | |
| BT029907.1 | - | 8 - 9 | 1084 | 89 | ||
Set Membership: |
|