Search
Search

CDC23| Gene Symbol: CDC23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080266.1 | NP_001073735.1 | 9 - 10 | 1012 | 108 | |
| GenBank mRNA | BC126843.1 | - | 9 - 10 | 1012 | 108 | |
Set Membership: |
|