Search
Search

POLD2| Gene Symbol: POLD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174428.3 | NP_776853.2 | 10 - 11 | 1327 | 61 | |
| GenBank mRNA | BC113315.1 | - | 9 - 10 | 1293 | 61 | |
| CK729402.1 | - | 1 - 2 | 263 | 61 | ||
| U21091.1 | - | 9 - 10 | 1310 | 61 | ||
Set Membership: |
|