Search
Search

IGFBP4| Gene Symbol: IGFBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174557.4 | NP_776982.1 | 3 - 4 | 884 | 75 | |
| GenBank mRNA | BC140688.1 | - | 3 - 4 | 885 | 75 | |
| S52770.1 | - | 3 - 4 | 848 | 75 | ||
Set Membership: |
|