Search
Search

PAG8| Gene Symbol: PAG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176619.3 | NP_788792.2 | 1 - 2 | 108 | 92 | |
| GenBank mRNA | AB099027.2 | - | 1 - 2 | 85 | 92 | |
| AF020510.2 | - | 1 - 2 | 64 | 92 | ||
| BC133476.1 | - | 1 - 2 | 109 | 92 | ||
Set Membership: |
|