Search
Search

LYVE1| Gene Symbol: LYVE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205815.2 | NP_991384.2 | 4 - 5 | 845 | 112 | |
| GenBank mRNA | AY372937.1 | - | 4 - 5 | 733 | 112 | |
| BC140639.1 | - | 4 - 5 | 845 | 112 | ||
Set Membership: |
|