Search
Search

C3H1orf50| Gene Symbol: C3H1orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034279.2 | NP_001029451.2 | 4 - 5 | 424 | 74 | |
| GenBank mRNA | BC102705.1 | - | 3 - 4 | 550 | 74 | |
| DT834744.1 | - | 4 - 5 | 426 | 74 | ||
Set Membership: |
|