Search
Search

IQGAP3| Gene Symbol: IQGAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098880.1 | NP_001092350.1 | 13 - 14 | 1469 | 65 | |
| GenBank mRNA | BC146242.1 | - | 13 - 14 | 1469 | 65 | |
Set Membership: |
|