Search
Search

NFATC2IP| Gene Symbol: NFATC2IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098883.2 | NP_001092353.1 | 6 - 7 | 1106 | 68 | |
| GenBank mRNA | BC142075.1 | - | 6 - 7 | 1107 | 68 | |
Set Membership: |
|