Search
Search

RPS19BP1| Gene Symbol: RPS19BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098916.2 | NP_001092386.1 | 3 - 4 | 347 | 57 | |
| GenBank mRNA | BC146267.1 | - | 3 - 4 | 348 | 57 | |
Set Membership: |
|