Search
Search

ALDH3A2| Gene Symbol: ALDH3A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101984.1 | NP_001095454.1 | 10 - 11 | 1514 | 126 | |
| GenBank mRNA | BC149984.1 | - | 10 - 11 | 1514 | 126 | |
Set Membership: |
|