Search
Search

PYROXD1| Gene Symbol: PYROXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105334.1 | NP_001098804.1 | 10 - 11 | 1137 | 68 | |
| GenBank mRNA | BC123726.1 | - | 10 - 11 | 1137 | 68 | |
Set Membership: |
|