Search
Search

KCTD19| Gene Symbol: KCTD19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105392.1 | NP_001098862.1 | 12 - 13 | 2260 | 74 | |
| GenBank mRNA | BC134572.1 | - | 12 - 13 | 2260 | 74 | |
Set Membership: |
|