Search
Search

MAT2A| Gene Symbol: MAT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101131.1 | NP_001094601.1 | 3 - 4 | 391 | 108 | |
| GenBank mRNA | BC111121.1 | - | 3 - 4 | 402 | 108 | |
| BT030708.1 | - | 3 - 4 | 391 | 108 | ||
Set Membership: |
|