Search
Search

C26H10orf82| Gene Symbol: C26H10orf82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101253.2 | NP_001094723.1 | 3 - 4 | 312 | 62 | |
| GenBank mRNA | BC149249.1 | - | 3 - 4 | 313 | 62 | |
Set Membership: |
|