Search
Search

TRAPPC11| Gene Symbol: TRAPPC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102197.1 | NP_001095667.1 | 12 - 13 | 1420 | 90 | |
| GenBank mRNA | BC147917.1 | - | 12 - 13 | 1420 | 90 | |
Set Membership: |
|