Search
Search

MYRFL| Gene Symbol: MYRFL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102323.1 | NP_001095793.1 | 19 - 20 | 2387 | 84 | |
| GenBank mRNA | BC148934.1 | - | 19 - 20 | 2387 | 84 | |
Set Membership: |
|