Search
Search

C14H8orf82| Gene Symbol: C14H8orf82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110184.1 | NP_001103654.1 | 2 - 3 | 230 | 75 | |
| GenBank mRNA | BC134700.1 | - | 2 - 3 | 230 | 75 | |
Set Membership: |
|