Search
Search

DPPA3| Gene Symbol: DPPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111108.2 | NP_001104578.1 | 4 - 5 | 552 | 76 | |
| NM_001111109.2 | NP_001104579.1 | 3 - 4 | 801 | 76 | ||
| GenBank mRNA | EF446904.1 | - | 3 - 4 | 801 | 76 | |
| EF446905.1 | - | 4 - 5 | 552 | 76 | ||
Set Membership: |
|