Search
Search

NELFA| Gene Symbol: NELFA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102353.1 | NP_001095823.1 | 6 - 7 | 919 | 65 | |
| GenBank mRNA | BC112828.1 | - | 6 - 7 | 854 | 65 | |
| BC149974.1 | - | 6 - 7 | 919 | 65 | ||
Set Membership: |
|