Search
Search

PRICKLE1| Gene Symbol: PRICKLE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102534.1 | NP_001096004.1 | 4 - 5 | 519 | 104 | |
| GenBank mRNA | BC140671.1 | - | 4 - 5 | 519 | 104 | |
Set Membership: |
|