Search
Search

CFAP73| Gene Symbol: CFAP73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144873.1 | NP_001138345.1 | 2 - 3 | 276 | 77 | |
| GenBank mRNA | EE906766.1 | - | 2 - 3 | 241 | 77 | |
| EV638271.1 | - | 2 - 3 | 279 | 77 | ||
Set Membership: |
|