Search
Search

CFAP73| Gene Symbol: CFAP73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144873.1 | NP_001138345.1 | 3 - 4 | 383 | 73 | |
| GenBank mRNA | EE906766.1 | - | 3 - 4 | 348 | 73 | |
| EV638271.1 | - | 3 - 4 | 386 | 73 | ||
Set Membership: |
|